missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human beta-Glucuronidase/GUSB Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 6144-GH-020

298.91 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18310528

  • £391.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to hydrolyze 4-methylumbelliferyl-beta-D-glucuronide. The specific activity is >4,000 pmol/min/μg, as measured under the described conditions.
Specifications

Specifications

AAA52561
Supplied as a 0.2 μm filtered solution in Tris and NaCl.
M.W. (Observed): 75-85 kDa, reducing conditions; M.W. (theoretical): 73 kDa
20 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
GUSB
Recombinant
beta-Glucuronidase/GUSB
Enzyme Activity
2990
beta-Glucuronidase/GUSB
Mouse myeloma cell line, NS0-derived human beta-Glucuronidase/GUSB protein Met1-Thr651, with a C-terminal 6-His tag
<1.0 EU / 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers