missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human BID (Caspase-8-cleaved) Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 882-B8-050

Product Code. 18177030

  • £370.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to induce cytochrome c release from isolated mouse liver mitochondria using the Rat/Mouse Cytochrome c Quantikine ELISA (Catalog # MCTC0 ) to quantify cytochrome c. The typical EC50 for cytochrome c releasing activity is <50 nM. The EC50 for the desired application should be determined. Uncleaved Recombinant Human BID (Catalog # 846-BD ) is available.
Specifications

Specifications

P55957
Supplied as a 0.2 μm filtered solution in HEPES-KOH and KCl.
M.W. (Observed): 9 kDa &14 kDa, reducing conditions; M.W. (theoretical): 7 kDa (N-terminal fragment) & 15 kDa (C-terminal fragment)
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Enzyme Activity
637
BID Caspase-8-cleaved
E. coli-derived human BID protein Met1-Asp195, protein was purified, cleaved by Caspase-8 and further purified
BID
Recombinant
BID
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.