missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-4 Protein
Click to view available options
Quantity:
1 mg
10 μg
50 μg
500 μg
Formulation:
Lyophilized from a 0.2 um filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Unit Size:
10µg
1mg
500µg
50µg
Description
Measured by its ability to induce BMP responsive SEAP reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 0.70-7.00 ng/mL. Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Binnerts, M.E. et al. (2004) Biochem. Biophys. Res. Commun. 315(2):272. The ED50 for this effect is 3.00-30.0 ng/mL.
Specifications
Specifications
| Accession Number | Q53XC5 |
| Characterization | Bioactivity |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
| Gene ID (Entrez) | 652 |
| Molecular Weight (g/mol) | M.W. (Observed): 19-24 kDa, reducing conditions 35-41 kDa, non-reducing conditions |
| Name | BMP-4 |
| Quantity | 1 mg |
| Source | Mouse myeloma cell line,NS0-derived human BMP-4 protein Ser293-Arg408 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction