missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-6 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£576.00
Specifications
Gene ID (Entrez) | 654 |
---|---|
Molecular Weight (g/mol) | 15 kDa (monomer) |
Name | BMP-6 |
Quantity | 20 μg |
Source | Mouse myeloma cell line,NS0-derived human BMP-6 protein Gln382-His513 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18397477
|
R&D Systems™
507-BP-020/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
£576.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18304817
|
R&D Systems™
507-BP-020 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
£576.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human BMP-6 Protein is derived from NS0. The Recombinant Human BMP-6 Protein has been validated for the following applications: Bioactivity.Specifications
654 | |
BMP-6 | |
Mouse myeloma cell line,NS0-derived human BMP-6 protein Gln382-His513 | |
Disulfide-linked homodimer | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
15 kDa (monomer) | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
BMP6, BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, VGR1, VGR-1 | |
Recombinant |