missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human BMP-7 Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP2-34933-10ug
139.20 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
P18075, P18075, P18075 | |
Western Blot, ELISA, PAGE | |
655 | |
10 μg | |
Less than 1 EU/ug of BMP-7 as determined by LAL method. | |
BMP7 | |
1. Molecular standard (Western, ELISA) in studying secreted BMP-7; 2. Preparing antibodies for BMP-7 monomer; 3. Molecule standard in detecting secreted BMP-7 in reduced SDS-PAGE. | |
>95% by SDS-PAGE and HPLC |
LYOPH | |
Lyophilized from a 0.2 um filtered concentrated solution in 30 % acetonitrile, 0.1 % TFA. | |
15.7 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1OP1, Osteogenic protein 1 | |
Recommended to centrifuge prior to opening. Reconstitute in 10 mM HAc to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. | |
Biologically Active Proteins, Cytokine Research | |
BMP-7 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human BMP-7 Protein > 10 μg
Spot an opportunity for improvement?Share a Content Correction