missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BNIP1 Protein

A cDNA sequence encoding the BNIP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11174-5ug

 View more versions of this product

Product Code. 15956378

  • £99.15 / 5µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

BNIP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAAPQDV HVRICNQEIV KFDLEVKALI QDIRDCSGPL SALTELNTKV KEKFQQLRHR IQDLEQLAKE QDKESEKQLL LQEVENHKKQ MLSNQASWRK ANLTCKIAID NLEKAELLQG GDLLRQRKTT KESLAQTSST ITESLMGISR MMAQQVQQSE EAMQSLVTSS RTILDANEEF KSMSGTIQLG RKLITKYNRR EL
Greater than 95% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
BNIP1 protein solution (1.0 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers