missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bone morphogenetic protein 1
A DNA sequence encoding the Homo sapiens (Human) Bone morphogenetic protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5722-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_001190.1 | |
Bone morphogenetic protein 1 | |
Research Use Only | |
BMP1 | |
Recombinant Protein | |
E. coli | |
AATSRPERVW PDGVIPFVIG GNFTGSQRAV FRQAMRHWEK HTCVTFLERT DEDSYIVFTY RPCGCCSYVG RRGGGPQAIS IGKNCDKFGI VVHELGHVVG FWHEHTRPDR DRHVSIVREN IQPGQEYNFL KMEPQEVESL GETYDFDSIM HYARNTFSRG IFLDTIVPKY EVNGVKPPIG QRTRLSKGDI AQARKLYKCP ACGETLQDST GNFSSPEYPN GYSAHMHCVW RISVTPGEKI ILNFTSLDLY RSRLCWYDYV EVRDGFWRKA PLRGRFCGSK LPEPIVSTDS RLWVEFRSSS NWVGKGFFAV YEAICGGDVK KDYGHIQSPN YPDDYRPSKV CIWRIQVSEG FHVGLTFQSF EIERHDSCAY DYLEVRDGHS ESSTLIGRYC GYEKPDDIKS TSSRLWLKFV SDGSINKAGF AVNFFKEVDE CSRPNRGGCE QRCLNTLGSY KCSCDPGYEL APDKRRCEAA CGGFLTKLNG SITSPGWPKE YPPNKNCIWQ LVAPTQYRIS LQFDFFETEG NDVCKYDFVE VRSGLTADSK LHGKFCGSEK PEVITSQYNN MRVEFKSDNT VSKKGFKAHF FSDKDECSKD NGGCQQDCVN TFGSYECQCR SGFVLHDNKH DCKEAGCDHK VTSTSGTITS PNWPDKYPSK KECTWAISST PGHRVKLTFM EMDIESQPEC AYDHLEVFDG RDAKAPVLGR FCGSKKPEPV LATGSRMFLR FYSDNSVQRK GFQASHATEC GGQVRADVKT KDLYSHAQFG DNNYPGGVDC EWVIVAEEGY GVELVFQTFE VEEETDCGYD YMELFDGYDS TAPRLGRYCG SGPPEEVYSA GDSVLVKFHS DDTITKKGFH LRYTSTKFQD TLHSRK | |
Greater than 90% as determined by SDS-PAGE. |
649 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |