missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bruton Tyrosine Kinase / BTK Kinase Protein
A DNA sequence encoding the Homo sapiens (Human) Bruton Tyrosine Kinase / BTK Kinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9013-ye-200ug
This item is not returnable.
View return policy
Specifications
NP_000052.1 | |
Bruton Tyrosine Kinase / BTK Kinase Protein | |
Research Use Only | |
Btk | |
Recombinant Protein | |
Yeast | |
AAVILESIFL KRSQQKKKTS PLNFKKRLFL LTVHKLSYYE YDFERGRRGS KKGSIDVEKI TCVETVVPEK NPPPERQIPR RGEESSEMEQ ISIIERFPYP FQVVYDEGPL YVFSPTEELR KRWIHQLKNV IRYNSDLVQK YHPCFWIDGQ YLCCSQTAKN AMGCQILENR NGSLKPGSSH RKTKKPLPPT PEEDQILKKP LPPEPAAAPV STSELKKVVA LYDYMPMNAN DLQLRKGDEY FILEESNLPW WRARDKNGQE GYIPSNYVTE AEDSIEMYEW YSKHMTRSQA EQLLKQEGKE GGFIVRDSSK AGKYTVSVFA KSTGDPQGVI RHYVVCSTPQ SQYYLAEKHL FSTIPELINY HQHNSAGLIS RLKYPVSQQN KNAPSTAGLG YGSWEIDPKD LTFLKELGTG QFGVVKYGKW RGQYDVAIKM IKEGSMSEDE FIEEAKVMMN LSHEKLVQLY GVCTKQRPIF IITEYMANGC LLNYLREMRH RFQTQQLLEM CKDVCEAMEY LESKQFLHRD LAARNCLVND QGVVKVSDFG LSRYVLDDEY TSSVGSKFPV RWSPPEVLMY SKFSSKSDIW AFGVLMWEIY SLGKMPYERF TNSETAEHIA QGLRLYRPHL ASEKVYTIMY SCWHEKADER PTFKILLSNI LDVMDEES | |
Greater than 90% as determined by SDS-PAGE. |
695 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |