missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BTN3A2 Protein
A DNA sequence encoding the Homo sapiens (Human) BTN3A2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5730-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001184175.1 | |
BTN3A2 Protein | |
Research Use Only | |
BTN3A2 | |
Recombinant Protein | |
E. coli | |
QFSVLGPSGP ILAMVGEDAD LPCHLFPTMS AETMELKWVS SSLRQVVNVY ADGKEVEDRQ SAPYRGRTSI LRDGITAGKA ALRIHNVTAS DSGKYLCYFQ DGDFYEKALV ELKVAALGSN LHVEVKGYED GGIHLECRST GWYPQPQIQW SNAKGENIPA VEAPVVADGV GLYEVAASVI MRGGSGEGVS CIIRNSLLGL EKTASISIAD PFFRSAQPW | |
Greater than 90% as determined by SDS-PAGE. |
11118 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |