missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C17ORF49 Protein
A cDNA sequence encoding the C17ORF49 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | C17ORF49 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976538
|
enQuireBio™
QP11225-2ug |
2 μg |
£99.15
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15956538
|
enQuireBio™
QP11225-10ug |
10 μg |
£155.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15966538
|
enQuireBio™
QP11225-1mg |
1 mg |
£5730.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
C17ORF49 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The C17ORF49 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTSASTK VGEIFSAAGA AFTKLGELTM QLHPVADSSP AGAKWTETEI EMLRAAVKRF GDDLNHISCV IKERTVAQIK ATVKRKVYED SGIPLPAESP KKGPKKVASG VLSPPPAAPP PSSSSVPEAG GPPIKKQKAD VTLSALNDSD ANSDVVDIEG LGETPPAKKL NFDQA | |
Greater than 90% as determined by SDS-PAGE. |