missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C9ORF103 Protein
A cDNA sequence encoding the C9ORF103 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11244-10ug
This item is not returnable.
View return policy
Specifications
C9ORF103 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAAPGA LLVMGVSGSG KSTVGALLAS ELGWKFYDAD DYHPEENRRK MGKGIPLNDQ DRIPWLCNLH DILLRDVASG QRVVLACSAL KKTYRDILTQ GKDGVALKCE ESGKEAKQAE MQLLVVHLSG SFEVISGRLL KREGHFMPPE LLQSQFETL PPAAPENFIQ ISVDKNVSEI IATIMETLKM K | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The C9ORF103 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol. |