missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CA125/MUC16 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand: R&D Systems™ 5609-MU-050
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human CA125/MUC16 Protein is derived from CHO. The Recombinant Human CA125/MUC16 Protein has been validated for the following applications: Binding Activity.Specifications
NP_078966.2 | |
94025 | |
CA125/MUC16 | |
Chinese Hamster Ovary cell line,CHO-derived human CA125/MUC16 protein Met13360-Gln14347 (Met13472Thr & Gln13957Lys) with a C-terminal 6-His tag | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Lyophilized from a 0.2μm filtered solution in PBS. | |
110 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |