missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calpain-1 catalytic subunit Protein
A DNA sequence encoding the Homo sapiens (Human) Calpain-1 catalytic subunit, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5754-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001185797.1 | |
Calpain-1 catalytic subunit Protein | |
Research Use Only | |
Capn1 | |
Recombinant Protein | |
E. coli | |
MSEEIITPVY CTGVSAQVQK QRARELGLGR HENAIKYLGQ DYEQLRVRCL QSGTLFRDEA FPPVPQSLGY KDLGPNSSKT YGIKWKRPTE LLSNPQFIVD GATRTDICQG ALGDCWLLAA IASLTLNDTL LHRVVPHGQS FQNGYAGIFH FQLWQFGEWV DVVVDDLLPI KDGKLVFVHS AEGNEFWSAL LEKAYAKVNG SYEALSGGST SEGFEDFTGG VTEWYELRKA PSDLYQIILK ALERGSLLGC SIDISSVLDM EAITFKKLVK GHAYSVTGAK QVNYRGQVVS LIRMRNPWGE VEWTGAWSDS SSEWNNVDPY ERDQLRVKME DGEFWMSFRD FMREFTRLEI CNLTPDALKS RTIRKWNTTL YEGTWRRGST AGGCRNYPAT FWVNPQFKIR LDETDDPDDY GDRESGCSFV LALMQKHRRR ERRFGRDMET IGFAVYEVPP ELVGQPAVHL KRDFFLANAS RARSEQFINL REVSTRFRLP PGEYVVVPST FEPNKEGDFV LRFFSEKSAG TVELDDQIQA NLPDEQVLSE EEIDENFKAL FRQLAGEDME ISVKELRTIL NRIISKHKDL RTKGFSLESC RSMVNLMDRD GNGKLGLVEF NILWNRIRNY LSIFRKFDLD KSGSMSAYEM RMAIESAGFK LNKKLYELII TRYSEPDLAV DFDNFVCCLV RLETMFRFFK TLDTDLDGVV TFDLFKWLQL TMFA | |
Greater than 90% as determined by SDS-PAGE. |
823 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |