missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Calumenin Protein

A cDNA sequence encoding the Calumenin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10543-1mg

 View more versions of this product

Product Code. 15944198

  • £5730.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Calumenin Protein
Research Use Only
Recombinant Protein
E. coli
MKPTEKKDRV HHEPQLSDKV HNDAQSFDYD HDAFLGAEEA KTFDQLTPEE SKERLGKIVS KIDGDKDGFV TVDELKDWIK FAQKRWIYED VERQWKGHDL NEDGLVSWEE YKNATYGYVL DDPDPDDGFN YKQMMVRDER RFKMADKDGD LIATKEEFTA FLHPEEYDYM KDIVVQETME DIDKNADGFI DLEEYIGDMY SHDGNTDEPE WVKTEREQFV EFRDKNRDGK MDKEETKDWI LPSDYDHAEA EARHLVYESD QNKDGKLTKE EIVDKYDLFV GSQATDFGEA LVRHDEF
Greater than 90.0% as determined by SDS-PAGE.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Human
Untagged
The Calumenin solution (0.5 mg/1ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers