missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calumenin Protein
A cDNA sequence encoding the Calumenin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10543-2ug
This item is not returnable.
View return policy
Specifications
Calumenin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKPTEKKDRV HHEPQLSDKV HNDAQSFDYD HDAFLGAEEA KTFDQLTPEE SKERLGKIVS KIDGDKDGFV TVDELKDWIK FAQKRWIYED VERQWKGHDL NEDGLVSWEE YKNATYGYVL DDPDPDDGFN YKQMMVRDER RFKMADKDGD LIATKEEFTA FLHPEEYDYM KDIVVQETME DIDKNADGFI DLEEYIGDMY SHDGNTDEPE WVKTEREQFV EFRDKNRDGK MDKEETKDWI LPSDYDHAEA EARHLVYESD QNKDGKLTKE EIVDKYDLFV GSQATDFGEA LVRHDEF | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human | |
Untagged | |
The Calumenin solution (0.5 mg/1ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |