missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Carbonic Anhydrase VIII / CA8 Protein
A DNA sequence encoding the Homo sapiens (Human) Carbonic Anhydrase VIII / CA8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5744-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001308766.1 | |
Carbonic Anhydrase VIII / CA8 Protein | |
Research Use Only | |
CA8 | |
Recombinant Protein | |
E. coli | |
MADLSFIEDT VAFPEKEEDE EEEEEGVEWG YEEGVEWGLV FPDANGEYQS PINLNSREAR YDPSLLDVRL SPNYVVCRDC EVTNDGHTIQ VILKSKSVLS GGPLPQGHEF ELYEVRFHWG RENQRGSEHT VNFKAFPMEL HLIHWNSTLF GSIDEAVGKP HGIAIIALFV QIGKEHVGLK AVTEILQDIQ YKGKSKTIPC FNPNTLLPDP LLRDYWVYEG SLTIPPCSEG VTWILFRYPL TISQLQIEEF RRLRTHVKGA ELVEGCDGIL GDNFRPTQPL SDRVIRAAFQ | |
Greater than 80% as determined by SDS-PAGE. |
767 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |