missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Cardiotrophin-1/CT-1 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 612-CD-050/CF

Additional Details : Weight : 0.00970kg

Product Code. 18762164

  • £725.68 / 50µg
Estimated Shipment: 05-08-2024
to see stock.

Description

Description

The Recombinant Human Cardiotrophin-1 (CT-1) Protein is derived from E. coli. The Recombinant Human Cardiotrophin-1 (CT-1) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Acetonitrile and TFA.
21 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening.
Recombinant
1489
Cardiotrophin-1/CT-1
E. coli-derived human Cardiotrophin-1/CT-1 protein Ser2-Ala201
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers