missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Cathepsin O GST (N-Term) Protein

Recombinant Protein

Brand:  Novus Biologicals™ H00001519-P01-10ug

 View more versions of this product

Product Code. 18317347

  • £483.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications

Specifications

Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification
1519
10 μg
cathepsin O, CTSO1, EC 3.4.22.42
>80% by SDS-PAGE and Coomassie blue staining
50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
M.W. (Theoretical): 58.52 kDa
Store at -80C. Avoid freeze-thaw cycles.
CTSO
Cathepsin O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.