missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Caveolin-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Caveolin-3, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001225.1 |
---|---|
Gene ID (Entrez) | 859 |
Name | Caveolin-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15947609
|
enQuireBio™
QP5766-ec-10ug |
10 μg | |||||||
15987609
|
enQuireBio™
QP5766-ec-50ug |
50 μg | |||||||
15937609
|
enQuireBio™
QP5766-ec-100ug |
100 μg |
£384.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15967609
|
enQuireBio™
QP5766-ec-200ug |
200 μg | |||||||
15977609
|
enQuireBio™
QP5766-ec-500ug |
500 μg | |||||||
15957609
|
enQuireBio™
QP5766-ec-1mg |
1 mg |
£1735.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001225.1 | |
Caveolin-3 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
859 | |
Research Use Only | |
CAV3 | |
Recombinant Protein | |
E. coli | |
MMAEEHTDLE AQIVKDIHCK EIDLVNRDPK NINEDIVKVD FEDVIAEPVG TYSFDGVWKV SYTTFTVSKY WCYRLLSTLL GVPLALLWGF LFACISFCHI WAVVPCIKSY LIEIQCISHI YSLCIRTFCN PLFAALGQVC SSIKVVLRKE V | |
Greater than 80% as determined by SDS-PAGE. |