Learn More
R&D Systems™ Recombinant Human CCL2/MCP-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 279-MC-001MG/CF
 View more versions of this product
Product Code. 18714987
  • £3429.72 / 1mg
Explore more special offers
Description
Description
The Recombinant Human CCL2/MCP-1 Protein is derived from E. coli. The Recombinant Human CCL2/MCP-1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
8.7 kDa
1mg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
6347
CCL2/JE/MCP-1
E. coli-derived human CCL2/JE/MCP-1 protein Gln24-Thr99
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.