Learn More
R&D Systems™ Recombinant Human CCL4L1/MIP-1 beta Isoform LAG-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 3046-MB-025/CF
 View more versions of this product
Product Code. 18359247
  • £188.00 / 25µg
Explore more special offers
Description
Description
The Recombinant Human CCL4L1/MIP-1 beta Isoform LAG-1 is derived from E. coli. The Recombinant Human CCL4L1/MIP-1 beta Isoform LAG-1 has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
7.8 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
9560
CCL4L1/LAG-1
E. coli-derived human CCL4L1/LAG-1 protein Ala24-Asn92
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.