missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD18 Protein
A cDNA sequence encoding the CD18 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Gene ID (Entrez) | 3689 |
---|---|
Name | CD18 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | ITGB2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966788
|
enQuireBio™
QP11307-2ug |
2 μg |
£99.15
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15946788
|
enQuireBio™
QP11307-10ug |
10 μg |
£155.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15956788
|
enQuireBio™
QP11307-1mg |
1 mg |
£5730.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
3689 | |
Research Use Only | |
ITGB2 | |
Human | |
His | |
CD18 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
CD18 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
QECTKFKVSS CRECIESGPG CTWCQKLNFT GPGDPDSIRC DTRPQLLMRG CAADDIMDPT SLAETQEDHN GGQKQLSPQK VTLYLRPGQA AAFNVTFRRA KGYPIDLYYL MDLSYSMLDD LRNVKKLGGD LLRALNEITE SGRIGFGSFV DKTVLPFVNT HPDKLRNPCP NKEKECQPPF AFRHVLKLTN NSNQFQTEVG KQLISGNLDA PEGGLDAMMQ VAACPEEIGW RNVTRLLVFA TDDGFHFAGD GKLGAILTPN DGRCHLEDNL YKRSNEFDYP SVGQLAHKLA ENNIQPIFAV TSRMVKTYEK LTEIIPKSAV GELSEDSSNV VQLIKNAYNK LSSRVFLDHN ALPDTLKVTY DSFCSNGVTH RNQPRGDCDG VQINVPITFQ VKVTATECIQ EQSFVIRALG FTDIVTVQVL PQCECRCRDQ SRDRSLCHGK GFLECGICRC DTGYIGKNCE CQTQGRSSQE LEGSCRKDNN SIICSGLGDC VCGQCLCHTS DVPGKLIYGQ YCECDTINCE RYNGQVCGGP GRGLCFCGKC RCHPGFEGSA CQCERTTEGC LNPRRVECSG RGRCRCNVCE CHSGYQLPLC QECPGCPSPC GKYISCAECL KFEKGPFGKN CSAACPGLQL SNNPVKGRTC KERDSEGCWV AYTLEQQDGM DRYLIYVDES RECVAGPNLE HHHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |