missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD1E Protein
A cDNA sequence encoding the CD1E was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 913 |
---|---|
Name | CD1E Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD1E |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986798
|
enQuireBio™
QP11311-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15976798
|
enQuireBio™
QP11311-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15966798
|
enQuireBio™
QP11311-1mg |
1 mg |
£2975.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
913 | |
Research Use Only | |
CD1E | |
Human | |
His | |
CD1E protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
CD1E Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEEQLSFR MLQTSSFANH SWAHSEGSGW LGDLQTHGWD TVLGTIRFLK PWSHGNFSKQ ELKNLQSLFQ LYFHSFIQIV QASAGQFQLE YPFEIQILAG CRMNAPQIFL NMAYQGSDFL SFQGISWEPS PGAGIRAQNI CKVLNRYLDI KEILQSLLGH TCPRFLAGLM EAGESELKRK VKPEAWLSCG PSPGPGRLQL VCHVSGFYPK PVWVMWMRGE QEQRGTQRGD VLPNADETWY LRATLDVAAG EAAGLSCRVK HSSLGGHDLI IHWGGYS | |
Greater than 85.0% as determined by SDS-PAGE. |