missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD276 Protein
A cDNA sequence encoding the CD276 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11321-10ug
This item is not returnable.
View return policy
Specifications
80381 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
LEVQVPEDPV VALVGTDATL CCSFSPEPGF SLAQLNLIWQ LTDTKQLVHS FAEGQDQGSA YANRTALFPD LLAQGNASLR LQRVRVADEG SFTCFVSIRD FGSAAVSLQV AAPYSKPSMT LEPNKDLRPG DTVTITCSSY QGYPEAEVFW QDGQGVPLTG NVTTSQMANE QGLFDVHSIL RVVLGANGTY SCLVRNPVLQ QDAHSSVTIT PQRSPTGAVE VQVPEDPVVA LVGTDATLRC SFSPEPGFSL AQLNLIWQLT DTKQLVHSFT EGRDQGSAYA NRTALFPDLL AQGNASLRLQ RVRVADEGSF TCFVSIRDFG SAAVSLQVAA PYSKPSMTLE PNKDLRPGDT VTITCSSYRG YPEAEVFWQD GQGVPLTGNV TTSQMANEQG LFDVHSVLRV VLGANGTYSC LVRNPVLQQD AHGSVTITGQ PMTFPPEALE HHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
CD276 Protein | |
Research Use Only | |
CD276 | |
Human | |
His | |
CD276 protein solution (0.5 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol. |