missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD34 Protein
A cDNA sequence encoding the CD34 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3305.00
Specifications
Gene ID (Entrez) | 947 |
---|---|
Name | CD34 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD34 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966878
|
enQuireBio™
QP11328-HIS-2ug |
2 μg |
£99.15
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15976878
|
enQuireBio™
QP11328-HIS-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15946878
|
enQuireBio™
QP11328-HIS-10ug |
10 μg |
£155.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15956878
|
enQuireBio™
QP11328-HIS-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15986878
|
enQuireBio™
QP11328-HIS-EC-1mg |
1 mg |
£3305.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
947 | |
Research Use Only | |
CD34 | |
Human | |
His | |
CD34 in 50mM Tris-HCl, pH 7.5, 10mM L-glutathione (reduced). |
CD34 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MLVRRGARAG PRMPRGWTAL CLLSLLPSGF MSLDNNGTAT PELPTQGTFS NVSTNVSYQE TTTPSTLGST SLHPVSQHGN EATTNITETT VKFTSTSVIT SVYGNTNSSV QSQTSVISTV FTTPANVSTP ETTLKPSLSP GNVSDLSTTS TSLATSPTKP YTSSSPILSD IKAEIKCSGI REVKLTQGIC LEQNKTSSCA EFKKDRGEGL ARVLCGEEQA DADAGAQVCS LLLAQSEVRP QCLLLVLANR TEISSKLQLM KKHQSDLKKL GILDFTEQDV ASHQSYSQKT LIALVTSGAL LAVLGITGYF LMNRRSWSPT GERLGEDPYY TENGGGQGYS SGPGTSPEAQ GKASVNRGAQ ENGTGQATSR NGHSARQHVV ADTEL |