missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD46 Protein
A cDNA sequence encoding the CD46 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11335-HIS-2ug
This item is not returnable.
View return policy
Specifications
4179 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSCEEPPTF EAMELIGKPK PYYEIGERVD YKCKKGYFYI PPLATHTICD RNHTWLPVSD DACYRETCPY IRDPLNGQAV PANGTYEFGY QMHFICNEGY YLIGEEILYC ELKGSVAIWS GKPPICEKVL CTPPPKIKNG KHTFSEVEVF EYLDAVTYSC DPAPGPDPFS LIGESTIYCG DNSVWSRAAP ECKVVKCRFP VVENGKQISG FGKKFYYKAT VMFECDKGFY LDGSDTIVCD SNSTWDPPVP KCLKGPRPTY KPPVSNYPGY PKPEEGILDS LD | |
Greater than 85% as determined by SDS-PAGE. |
CD46 Protein | |
Research Use Only | |
CD46 | |
Human | |
His | |
CD46 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 20% glycerol. |