missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD81 / TAPA-1 Protein
A DNA sequence encoding the Homo sapiens (Human) CD81 / TAPA-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5797-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001284578.1 | |
CD81 / TAPA-1 Protein | |
Research Use Only | |
Cd81 | |
Recombinant Protein | |
Yeast | |
FVNKDQIAKD VKQFYDQALQ QAVVDDDANN AKAVVKTFHE TLDCCGSSTL TALTTSVLKN NLCPSGSNII SNLFKEDCHQ KIDDLFSGK | |
Greater than 90% as determined by SDS-PAGE. |
975 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |