missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CENPA Protein
A cDNA sequence encoding the CENPA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11387-20ug
This item is not returnable.
View return policy
Specifications
CENPA Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MGPRRRSRKP EAPRRRSPSP TPTPGPSRRG PSLGASSHQH SRRRQGWLKE IRKLQKSTHL LIRKLPFSRL AREICVKFTR GVDFNWQAQA LLALQEAAEA FLVHLFEDAY LLTLHAGRVT LFPKDVQLAR RIRGLEEGLG | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CENPA is supplied in 20mM HEPES buffer pH 8.0, 100mM sodium chloride, and 6M Urea. |