missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Centromere protein 1
A DNA sequence encoding the Homo sapiens (Human) Centromere protein K, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5819-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001253967.1 | |
Centromere protein 1 | |
Research Use Only | |
CENPK | |
Recombinant Protein | |
E. coli | |
MNQEDLDPDS TTDVGDVTNT EEELIRECEE MWKDMEECQN KLSLIGTETL TDSNAQLSLL IMQVKCLTAE LSQWQKKTPE TIPLTEDVLI TLGKEEFQKL RQDLEMVLST KESKNEKLKE DLEREQRWLD EQQQIMESLN VLHSELKNKV ETFSESRIFN ELKTKMLNIK EYKEKLLSTL GEFLEDHFPL PDRSVKKKKK NIQESSVNLI TLHEMLEILI NRLFDVPHDP YVKISDSFWP PYVELLLRNG IALRHPEDPT RIRLEAFHQ | |
Greater than 80% as determined by SDS-PAGE. |
64105 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |