missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Centromere protein 1

A DNA sequence encoding the Homo sapiens (Human) Centromere protein K, was expressed in the hosts and tags indicated.

£223.39 - £2480.00

Specifications

Accession Number NP_001253967.1
Gene ID (Entrez) 64105
Name Centromere protein 1
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15957989
View Documents Promotion Details
enQuireBio™
QP5819-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15997989
View Documents Promotion Details
enQuireBio™
QP5819-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15947989
View Documents Promotion Details
enQuireBio™
QP5819-ec-100ug
100 μg
£560.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15977989
View Documents Promotion Details
enQuireBio™
QP5819-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15987989
View Documents Promotion Details
enQuireBio™
QP5819-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15967989
View Documents Promotion Details
enQuireBio™
QP5819-ec-1mg
1 mg
£2480.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001253967.1
Centromere protein 1
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
64105
Research Use Only
CENPK
Recombinant Protein
E. coli
MNQEDLDPDS TTDVGDVTNT EEELIRECEE MWKDMEECQN KLSLIGTETL TDSNAQLSLL IMQVKCLTAE LSQWQKKTPE TIPLTEDVLI TLGKEEFQKL RQDLEMVLST KESKNEKLKE DLEREQRWLD EQQQIMESLN VLHSELKNKV ETFSESRIFN ELKTKMLNIK EYKEKLLSTL GEFLEDHFPL PDRSVKKKKK NIQESSVNLI TLHEMLEILI NRLFDVPHDP YVKISDSFWP PYVELLLRNG IALRHPEDPT RIRLEAFHQ
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications