missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Charged multivesicular body protein 2b Protein
A DNA sequence encoding the Homo sapiens (Human) Charged multivesicular body protein 2b, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8250-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_001231573.1 | |
Charged multivesicular body protein 2b Protein | |
Research Use Only | |
CHMP2B | |
Recombinant Protein | |
E. coli | |
ASLFKKKTVD DVIKEQNREL RGTQRAIIRD RAALEKQEKQ LELEIKKMAK IGNKEACKVL AKQLVHLRKQ KTRTFAVSSK VTSMSTQTKV MNSQMKMAGA MSTTAKTMQA VNKKMDPQKT LQTMQNFQKE NMKMEMTEEM INDTLDDIFD GSDDEEESQD IVNQVLDEIG IEISGKMAKA PSAARSLPSA STSKATISDE EIERQLKALG VD | |
Greater than 80% as determined by SDS-PAGE. |
25978 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |