missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Charged multivesicular body protein 2b Protein
A DNA sequence encoding the Homo sapiens (Human) Charged multivesicular body protein 2b, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001231573.1 |
---|---|
Gene ID (Entrez) | 25978 |
Name | Charged multivesicular body protein 2b Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16073561
|
enQuireBio™
QP8250-ec-10ug |
10 μg | |||||||
16013571
|
enQuireBio™
QP8250-ec-50ug |
50 μg | |||||||
16063561
|
enQuireBio™
QP8250-ec-100ug |
100 μg |
£384.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16093561
|
enQuireBio™
QP8250-ec-200ug |
200 μg | |||||||
16003571
|
enQuireBio™
QP8250-ec-500ug |
500 μg | |||||||
16083561
|
enQuireBio™
QP8250-ec-1mg |
1 mg |
£1735.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001231573.1 | |
Charged multivesicular body protein 2b Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
25978 | |
Research Use Only | |
CHMP2B | |
Recombinant Protein | |
E. coli | |
ASLFKKKTVD DVIKEQNREL RGTQRAIIRD RAALEKQEKQ LELEIKKMAK IGNKEACKVL AKQLVHLRKQ KTRTFAVSSK VTSMSTQTKV MNSQMKMAGA MSTTAKTMQA VNKKMDPQKT LQTMQNFQKE NMKMEMTEEM INDTLDDIFD GSDDEEESQD IVNQVLDEIG IEISGKMAKA PSAARSLPSA STSKATISDE EIERQLKALG VD | |
Greater than 80% as determined by SDS-PAGE. |