missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CHRNA6 Protein
A cDNA sequence encoding the CHRNA6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11426-5ug
This item is not returnable.
View return policy
Specifications
CHRNA6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSKGCV GCATEERLFH KLFSHYNQFI RPVENVSDPV TVHFEVAITQ LANVDEVNQI METNLWLRHI WNDYKLRWDP MEYDGIETLR VPADKIWKPD IVLYNNAVGD FQVEGKTKAL LKYNGMITWT PPAIFKSSCP MDITFFPFDH QNCSLKFGSW TYDKAEIDLL IIGSKVDMND FWENSEWEII DASGYKHDIK YNCCEEIYTD ITYSFYIRRL | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CHRNA6 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |