missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CINP Protein

A DNA sequence encoding the Homo sapiens (Human) CINP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8044-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16082341

  • £384.00 / 100µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001306975.1
CINP Protein
Research Use Only
CINP
Recombinant Protein
E. coli
MEAKTLGTVT PRKPVLSVSA RKIKDNAADW HNLILKWETL NDAGFTTANN IANLKISLLN KDKIELDSSS PASKENEEKV CLEYNEELEK LCEELQATLD GLTKIQVKME KLSSTTKGIC ELENYHYGEE SKRPPLFHTW PTTHFYEVSH KLLEMYRKEL LLKRTVAKEL AHTGDPDLTL SYLSMWLHQP YVESDSRLHL ESMLLETGHR AL
Greater than 90% as determined by SDS-PAGE.
51550
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers