missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLEC4E Protein
A cDNA sequence encoding the CLEC4E was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | CLEC4E Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15967288
|
enQuireBio™
QP11445-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15957288
|
enQuireBio™
QP11445-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15947288
|
enQuireBio™
QP11445-1mg |
1 mg |
£2975.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CLEC4E Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CLEC4E protein solution (0.25 mg/ml) containing 20mM Tris 8.0 and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MRCVVTFRIF QTCDEKKFQL PENFTELSCY NYGSGSVKNC CPLNWEYFQS SCYFFSTDTI SWALSLKNCS AMGAHLVVIN SQEEQEFLSY KKPKMREFFI GLSDQVVEGQ WQWVDGTPLT KSLSFWDVGE PNNIATLEDC ATMRDSSNPR QNWNDVTCFL NYFRICEMVG INPLNKGKSL | |
Greater than 85.0% as determined by SDS-PAGE. |