missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Clusterin Protein
A DNA sequence encoding the Homo sapiens (Human) Clusterin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8529-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001822.3 | |
Clusterin Protein | |
Research Use Only | |
APOJ | |
Recombinant Protein | |
E. coli | |
DQTVSDNELQ EMSNQGSKYV NKEIQNAVNG VKQIKTLIEK TNEERKTLLS NLEEAKKKKE DALNETRESE TKLKELPGVC NETMMALWEE CKPCLKQTCM KFYARVCRSG SGLVGRQLEE FLNQSSPFYF WMNGDRIDSL LENDRQQTHM LDVMQDHFSR ASSIIDELFQ DRFFTREPQD TYHYLPFSLP HRRPHFFFPK SR | |
Greater than 90% as determined by SDS-PAGE. |
1191 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |