missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Clusterin Protein
A DNA sequence encoding the Homo sapiens (Human) Clusterin, was expressed in the hosts and tags indicated.
£145.46 - £1688.38
Specifications
Accession Number | NP_001822.3 |
---|---|
Gene ID (Entrez) | 1191 |
Name | Clusterin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16015121
|
enQuireBio™
QP8529-ec-10ug |
10 μg | |||||||
16055121
|
enQuireBio™
QP8529-ec-50ug |
50 μg | |||||||
16005121
|
enQuireBio™
QP8529-ec-100ug |
100 μg | |||||||
16035121
|
enQuireBio™
QP8529-ec-200ug |
200 μg | |||||||
16045121
|
enQuireBio™
QP8529-ec-500ug |
500 μg | |||||||
16025121
|
enQuireBio™
QP8529-ec-1mg |
1 mg | |||||||
Specifications
NP_001822.3 | |
Clusterin Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1191 | |
Research Use Only | |
APOJ | |
Recombinant Protein | |
E. coli | |
DQTVSDNELQ EMSNQGSKYV NKEIQNAVNG VKQIKTLIEK TNEERKTLLS NLEEAKKKKE DALNETRESE TKLKELPGVC NETMMALWEE CKPCLKQTCM KFYARVCRSG SGLVGRQLEE FLNQSSPFYF WMNGDRIDSL LENDRQQTHM LDVMQDHFSR ASSIIDELFQ DRFFTREPQD TYHYLPFSLP HRRPHFFFPK SR | |
Greater than 90% as determined by SDS-PAGE. |