missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CMC4 Protein
A cDNA sequence encoding the CMC4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11458-5ug
This item is not returnable.
View return policy
Specifications
CMC4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEFMPQKD PCQKQACEIQ KCLQANSYME SKCQAVIQEL RKCCAQYPKG RSVVCSGFEK EEEENLTRKS ASK | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CMC4 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 20% glycerol. |