missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-1(XII) chain Protein
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(XII) chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8023-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004361.3 | |
Collagen alpha-1(XII) chain Protein | |
Research Use Only | |
COL12A1 | |
Recombinant Protein | |
E. coli | |
DLVFLVDGSW SVGRNNFKYI LDFIAALVSA FDIGEEKTRV GVVQYSSDTR TEFNLNQYYQ RDELLAAIKK IPYKGGNTMT GDAIDYLVKN TFTESAGARV GFPKVAIIIT DGKSQDEVEI PARELRNVGV EVFSLGIKAA DAKELKQIAS TPSLNHVFNV ANFDAIVDIQ NEIISQV | |
Greater than 90% as determined by SDS-PAGE. |
1303 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |