missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Collagen alpha-1(XII) chain Protein

A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(XII) chain, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8023-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062211

  • £384.00 / 100µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_004361.3
Collagen alpha-1(XII) chain Protein
Research Use Only
COL12A1
Recombinant Protein
E. coli
DLVFLVDGSW SVGRNNFKYI LDFIAALVSA FDIGEEKTRV GVVQYSSDTR TEFNLNQYYQ RDELLAAIKK IPYKGGNTMT GDAIDYLVKN TFTESAGARV GFPKVAIIIT DGKSQDEVEI PARELRNVGV EVFSLGIKAA DAKELKQIAS TPSLNHVFNV ANFDAIVDIQ NEIISQV
Greater than 90% as determined by SDS-PAGE.
1303
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers