missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Complement C4-B Protein
A DNA sequence encoding the Homo sapiens (Human) Complement C4-B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6998-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001002029.3 | |
Complement C4-B Protein | |
Research Use Only | |
C4B | |
Recombinant Protein | |
E. coli | |
EAPKVVEEQE SRVHYTVCIW RNGKVGLSGM AIADVTLLSG FHALRADLEK LTSLSDRYVS HFETEGPHVL LYFDSVPTSR ECVGFEAVQE VPVGLVQPAS ATLYDYYNPE RRCSVFYGAP SKSRLLATLC SAEVCQCAEG KCPRQRRALE RGLQDEDGYR MKFACYYPRV EYGFQVKVLR EDSRAAFRLF ETKITQVLHF TKDVKAAANQ MRNFLVRASC RLRLEPGKEY LIMGLDGATY DLEGHPQYLL DSNSWIEEMP SERLCRSTRQ RAACAQLNDF LQEYGTQGCQ V | |
Greater than 90% as determined by SDS-PAGE. |
100293534 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction