missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COX4I1, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) COX4I1, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5854-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001305715.1 | |
COX4I1, mitochondrial Protein | |
Research Use Only | |
COX4I1 | |
Recombinant Protein | |
E. coli | |
AHESVVKSED FSLPAYMDRR DHPLPEVAHV KHLSASQKAL KEKEKASWSS LSMDEKVELY RIKFKESFAE MNRGSNEWKT VVGGAMFFIG FTALVIMWQK HYVYGPLPQS FDKEWVAKQT KRMLDMKVNP IQGLASKWDY EKNEWKK | |
Greater than 90% as determined by SDS-PAGE. |
1327 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |