missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CSNK2A1 / CK2A1 Protein
A DNA sequence encoding the Homo sapiens (Human) CSNK2A1 / CK2A1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5881-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001886.1 | |
CSNK2A1 / CK2A1 Protein | |
Research Use Only | |
CSNK2A1 | |
Recombinant Protein | |
E. coli | |
MSGPVPSRAR VYTDVNTHRP REYWDYESHV VEWGNQDDYQ LVRKLGRGKY SEVFEAINIT NNEKVVVKIL KPVKKKKIKR EIKILENLRG GPNIITLADI VKDPVSRTPA LVFEHVNNTD FKQLYQTLTD YDIRFYMYEI LKALDYCHSM GIMHRDVKPH NVMIDHEHRK LRLIDWGLAE FYHPGQEYNV RVASRYFKGP ELLVDYQMYD YSLDMWSLGC MLASMIFRKE PFFHGHDNYD QLVRIAKVLG TEDLYDYIDK YNIELDPRFN DILGRHSRKR WERFVHSENQ HLVSPEALDF LDKLLRYDHQ SRLTAREAME HPYFYTVVKD QARMGSSSMP GGSTPVSSAN MMSGISSVPT PSPLGPLAGS PVIAAANPLG MPVPAAAGAQ Q | |
Greater than 90% as determined by SDS-PAGE. |
1457 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |