missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CTHRC1 Protein
A cDNA sequence encoding the CTHRC1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11554-HIS-10ug
This item is not returnable.
View return policy
Specifications
CTHRC1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSEIPKGKQK AQLRQREVVD LYNGMCLQGP AGVPGRDGSP GANGIPGTPG IPGRDGFKGE KGECLRESFE ESWTPNYKQC SWSSLNYGID LGKIAECTFT KMRSNSALRV LFSGSLRLKC RNACCQRWYF TFNGAECSGP LPIEAIIYLD QGSPEMNSTI NIHRTSSVEG LCEGIGAGLV DVAIWVGTCS DYPKGDASTG WNSVSRIIIE ELPK | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CTHRC1 protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |