missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CTSL Protein

A cDNA sequence encoding the CTSL was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11561-5ug

 View more versions of this product

Product Code. 15957648

  • £99.15 / 5µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

1514
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSTLTFDHS LEAQWTKWKA MHNRLYGMNE EGWRRAVWEK NMKMIELHNQ EYREGKHSFT MAMNAFGDMT SEEFRQVMNG FQNRKPRKGK VFQEPLFYEA PRSVDWREKG YVTPVKNQGQ CGSCWAFSAT GALEGQMFRK TGRLISLSEQ NLVDCSGPQG NEGCNGGLMD YAFQYVQDNG GLDSEESYPY EATEESCKYN PKYSVANDTG FVDIPKQEKA LMKAVATVGP ISVAIDAGHE SFLFYKEGIY FEPDCSSEDM DHGVLVVGYG FESTESDNNK YWLVKNSWGE EWGMGGYVKM AKDRRNHCGI ASAASYPTV
Greater than 90% as determined by SDS-PAGE.
CTSL Protein
Research Use Only
CTSL
Human
His
CTSL protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers