missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CXCL8/IL-8 Protein
Recombinant Protein
£105.00 - £2205.00
Specifications
Accession Number | P10145, P10145, P10145 |
---|---|
Concentration | LYOPH |
For Use With (Application) | PAGE, Bioactivity |
Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
Gene ID (Entrez) | 3576 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18407701
|
Novus Biologicals™
NBP2-34905-1mg |
1 mg |
£2205.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18446011
|
Novus Biologicals™
NBP2-34905-100ug |
100 μg |
£579.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18442472
|
Novus Biologicals™
NBP2-34905-500ug |
500 μg |
£1270.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18417431
|
Novus Biologicals™
NBP2-34905-5ug |
5 μg |
£105.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fully biologically active when compared to standard. The ED50 as determined by a chemotaxis bioassay using human CXCR2 transfected mouse BaF3 cells is less than 2 ng/mL, corresponding to a specific activity of >5.0 x 10^5 IU/mg.Specifications
P10145, P10145, P10145 | |
PAGE, Bioactivity | |
3576 | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, IL-8/CXCL8, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. | |
>97% pure by SDS-PAGE and HPLC |
LYOPH | |
Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. | |
8.4 kDa | |
Less than 1 EU/ug of CXCL8/IL-8 as determined by LAL method. | |
CXCL8 | |
Angiogenesis, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
CXCL8/IL-8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human CXCL8/IL-8 Protein