missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cyclic GMP-AMP synthase Protein
A DNA sequence encoding the Homo sapiens (Human) Cyclic GMP-AMP synthase, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_612450.2 |
---|---|
Gene ID (Entrez) | 115004 |
Name | Cyclic GMP-AMP synthase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16001191
|
enQuireBio™
QP7853-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021191
|
enQuireBio™
QP7853-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011191
|
enQuireBio™
QP7853-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031191
|
enQuireBio™
QP7853-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041191
|
enQuireBio™
QP7853-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051191
|
enQuireBio™
QP7853-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_612450.2 | |
Cyclic GMP-AMP synthase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
115004 | |
Research Use Only | |
MB21D1 | |
Recombinant Protein | |
E. coli | |
GASKLRAVLE KLKLSRDDIS TAAGMVKGVV DHLLLRLKCD SAFRGVGLLN TGSYYEHVKI SAPNEFDVMF KLEVPRIQLE EYSNTRAYYF VKFKRNPKEN PLSQFLEGEI LSASKMLSKF RKIIKEEIND IKDTDVIMKR KRGGSPAVTL LISEKISVDI TLALESKSSW PASTQEGLRI QNWLSAKVRK QLRLKPFYLV PKHAKEGNGF QEETWRLSFS HIEKEILNNH GKSKTCCENK EEKCCRKDCL KLMKYLLEQL KERFKDKKHL DKFSSYHVKT AFFHVCTQNP QDSQWDRKDL GLCFDNCVTY FLQCLRTEKL ENYFIPEFNL FSSNLIDKRS KEFLTKQIEY ERNNEFPVFD EF | |
Greater than 90% as determined by SDS-PAGE. |