missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytochrome C Protein
A DNA sequence encoding the Homo sapiens (Human) Cytochrome C, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_061820.1 |
---|---|
Gene ID (Entrez) | 54205 |
Name | Cytochrome C Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948539
|
enQuireBio™
QP5900-ec-10ug |
10 μg | |||||||
15988539
|
enQuireBio™
QP5900-ec-50ug |
50 μg | |||||||
15938539
|
enQuireBio™
QP5900-ec-100ug |
100 μg |
£384.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15968539
|
enQuireBio™
QP5900-ec-200ug |
200 μg | |||||||
15978539
|
enQuireBio™
QP5900-ec-500ug |
500 μg | |||||||
15958539
|
enQuireBio™
QP5900-ec-1mg |
1 mg |
£1735.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_061820.1 | |
Cytochrome C Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
54205 | |
Research Use Only | |
CYCS | |
Recombinant Protein | |
E. coli | |
GDVEKGKKIF IMKCSQCHTV EKGGKHKTGP NLHGLFGRKT GQAPGYSYTA ANKNKGIIWG EDTLMEYLEN PKKYIPGTKM IFVGIKKKEE RADLIAYLKK ATNE | |
Greater than 90% as determined by SDS-PAGE. |