missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Cytosolic beta-Glucosidase/GBA3

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 5969-GH-010

Product Code. 18370058

  • £343.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to hydrolyze 4-methylumbelliferyl-beta-D-glucopyranoside. The specific activity is >1,500 pmol/min/μg, as measured under the described conditions.
Specifications

Specifications

Q9H227
Supplied as a 0.2 μm filtered solution in Tris, NaCl, DTT and Glycerol.
M.W. (Observed): 54 kDa, reducing conditions; M.W. (theoretical): 53 kDa & 41 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
GBA3
Recombinant
Cytosolic beta-Glucosidase/GBA3
Enzyme Activity
57733
Cytosolic beta-Glucosidase/GBA3
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Cytosolic beta-Glucosidase/GBA3 protein Thr13-Leu469 with a C-terminal 6-His tag
<1.0 EU / 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.