missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human D-amino-acid oxidase Protein
A DNA sequence encoding the Homo sapiens (Human) D-amino-acid oxidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5910-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001908.3 | |
D-amino-acid oxidase Protein | |
Research Use Only | |
DAO | |
Recombinant Protein | |
E. coli | |
MRVVVIGAGV IGLSTALCIH ERYHSVLQPL DIKVYADRFT PLTTTDVAAG LWQPYLSDPN NPQEADWSQQ TFDYLLSHVH SPNAENLGLF LISGYNLFHE AIPDPSWKDT VLGFRKLTPR ELDMFPDYGY GWFHTSLILE GKNYLQWLTE RLTERGVKFF QRKVESFEEV AREGADVIVN CTGVWAGALQ RDPLLQPGRG QIMKVDAPWM KHFILTHDPE RGIYNSPYII PGTQTVTLGG IFQLGNWSEL NNIQDHNTIW EGCCRLEPTL KNARIIGERT GFRPVRPQIR LEREQLRTGP SNTEVIHNYG HGGYGLTIHW GCALEAAKLF GRILEEKKLS RMPPSHL | |
Greater than 90% as determined by SDS-PAGE. |
1610 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |